Learn More
3,3-Dimethylpentane, 99%, Thermo Scientific Chemicals
£37.50 - £454.00
Chemical Identifiers
CAS | 562-49-2 |
---|---|
Molecular Formula | C7H16 |
Molecular Weight (g/mol) | 100.2 |
MDL Number | MFCD00009322 |
InChI Key | AEXMKKGTQYQZCS-UHFFFAOYSA-N |
PubChem CID | 11229 |
IUPAC Name | 3,3-dimethylpentane |
SMILES | CCC(C)(C)CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10275910
|
Thermo Scientific Acros
152660010 |
1 g |
£37.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10153680
|
Thermo Scientific Acros
152660050 |
5 g |
£124.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10279393
|
Thermo Scientific Acros
152660250 |
25 g |
£454.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
562-49-2 | |
100.2 | |
AEXMKKGTQYQZCS-UHFFFAOYSA-N | |
3,3-dimethylpentane |
C7H16 | |
MFCD00009322 | |
11229 | |
CCC(C)(C)CC |
Specifications
562-49-2 | |
100.0 | |
Colorless | |
86°C | |
Authentic | |
Glass Bottle | |
C2H5C(CH3)2C2H5 | |
MFCD00009322 | |
0.69 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether,miscible with acetone | |
CCC(C)(C)CC | |
100.2 | |
100.2 | |
Liquid |
98.5 | |
-135°C | |
0.6900g/mL | |
-6°C | |
98.5% min. (GC) | |
C7H16 | |
1.3900 to 1.392 | |
01,158 | |
pentane, 3,3-dimethyl, pentane,3,3-dimethyl, unii-r1rh0vbd2l, r1rh0vbd2l, 3,3,-dimethylpentane, acmc-1b14l, dsstox_cid_29275, dsstox_rid_83394, dsstox_gsid_49319, 3,3-dimethylpentane | |
AEXMKKGTQYQZCS-UHFFFAOYSA-N | |
3,3-dimethylpentane | |
11229 | |
99% | |
3, 3-Dimethylpentane |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-230-8
- 3,3-Dimethylpentane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P331-Do NOT induce vomiting.
RUO – Research Use Only