missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylpentane, 99%

Product Code. 10275910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10275910 1 g 1g
10153680 5 g 5g
10279393 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10275910 Supplier Thermo Scientific Acros Supplier No. 152660010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 562-49-2 | C7H16 | 100.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 562-49-2
Molecular Formula C7H16
Molecular Weight (g/mol) 100.2
MDL Number MFCD00009322
InChI Key AEXMKKGTQYQZCS-UHFFFAOYSA-N
PubChem CID 11229
IUPAC Name 3,3-dimethylpentane
SMILES CCC(C)(C)CC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -135°C
Color Colorless
Density 0.6900g/mL
Boiling Point 86°C
Flash Point -6°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula C2H5C(CH3)2C2H5
Refractive Index 1.3900 to 1.392
Quantity 1 g
Beilstein 01,158
Specific Gravity 0.69
Synonym pentane, 3,3-dimethyl, pentane,3,3-dimethyl, unii-r1rh0vbd2l, r1rh0vbd2l, 3,3,-dimethylpentane, acmc-1b14l, dsstox_cid_29275, dsstox_rid_83394, dsstox_gsid_49319, 3,3-dimethylpentane
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether,miscible with acetone
Formula Weight 100.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3, 3-Dimethylpentane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3,3-Dimethylpentane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.