missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-1-butene, 95%

Product Code. p-116359 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
This item is not returnable. View return policy

Product Code. 10245960

Brand: Thermo Scientific Acros 152722500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 558-37-2 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 558-37-2
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00008853
InChI Key PKXHXOTZMFCXSH-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1-butene, neohexene, 1-butene, 3,3-dimethyl, tert-butylethylene, tert-butylethene, tert-hexene, trimethylvinylmethane, 3,3-dimethylbutene, 2,2-dimethyl-3-butene, unii-01zb73d2kk
PubChem CID 11210
IUPAC Name 3,3-dimethylbut-1-ene
SMILES CC(C)(C)C=C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -115.0°C
Color Colorless
Density 0.6500g/mL
Boiling Point 41.0°C
Flash Point −28°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCH=CH2
Refractive Index 1.3750 to 1.3780
Quantity 250 mL
Beilstein 01, 217
Specific Gravity 0.65
Formula Weight 84.16
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-1-butene, 95%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3,3-Dimethyl-1-butene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.