accessibility menu, dialog, popup

UserName

CAS RN 558-37-2

IUPAC Name: 3,3-dimethylbut-1-ene
Synonyms: 3,3-dimethyl-1-butene
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: PKXHXOTZMFCXSH-UHFFFAOYSA-N
SMILES: CC(C)(C)C=C
Boiling Point 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)