missing translation for 'onlineSavingsMsg'
Learn More

3,3-Difluoropiperidine hydrochloride, 99%, Thermo Scientific™

Product Code. 11339735
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339735 250 mg 250mg
11329735 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339735 Supplier Thermo Scientific Alfa Aesar Supplier No. L19777.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496807-97-7
Molecular Formula C5H10ClF2N
Molecular Weight (g/mol) 157.589
MDL Number MFCD03452800
InChI Key LEHHIPIDKQVNEV-UHFFFAOYSA-N
Synonym 3,3-difluoropiperidine hydrochloride, 3,3-difluoropiperidine hcl, piperidine, 3,3-difluoro-, hydrochloride, 3,3-difluoropiperidine, chloride, pubchem10314, acmc-1an4u, 3,3-difluoropiperidine-hcl, 3,3-difluoro-piperidine hcl, 3,3-difluoro-piperidine hydrochloride, 3,3-bis fluoranyl piperidine hydrochloride
PubChem CID 2758349
IUPAC Name 3,3-difluoropiperidine;hydrochloride
SMILES C1CC(CNC1)(F)F.Cl

Specifications

Melting Point ∼240°C
Quantity 250 mg
Sensitivity Hygroscopic
Formula Weight 157.59
Percent Purity 99%
Chemical Name or Material 3,3-Difluoropiperidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.