CAS RN 496807-97-7
IUPAC Name:
3,3-difluoropiperidine hydrochloride
Synonyms:
3,3-difluoropiperidine hydrochloride
Molecular Weight (g/mol):
157.59
Molecular Formula:
C5H10ClF2N
InChi Key:
LEHHIPIDKQVNEV-UHFFFAOYSA-N
SMILES:
Cl.FC1(F)CCCNC1
1
–
1
of
1
results
1
3,3-Difluoropiperidine hydrochloride, 99%, Thermo Scientific™
CAS: 496807-97-7 Molecular Formula: C5H10ClF2N Molecular Weight (g/mol): 157.589 MDL Number: MFCD03452800 InChI Key: LEHHIPIDKQVNEV-UHFFFAOYSA-N Synonym: 3,3-difluoropiperidine hydrochloride,3,3-difluoropiperidine hcl,piperidine, 3,3-difluoro-, hydrochloride,3,3-difluoropiperidine, chloride,pubchem10314,acmc-1an4u,3,3-difluoropiperidine-hcl,3,3-difluoro-piperidine hcl,3,3-difluoro-piperidine hydrochloride,3,3-bis fluoranyl piperidine hydrochloride PubChem CID: 2758349 IUPAC Name: 3,3-difluoropiperidine;hydrochloride SMILES: C1CC(CNC1)(F)F.Cl