missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 11327669
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327669 1 g 1g
11337669 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327669 Supplier Thermo Scientific Alfa Aesar Supplier No. H28340.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073353-64-6
Molecular Formula C16H24BNO4
Molecular Weight (g/mol) 305.18
MDL Number MFCD09266187
InChI Key GNJHHGGRMLSKNG-UHFFFAOYSA-N
Synonym n-2-methoxyethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 3-2-methoxyethylaminocarbonyl benzeneboronic acid pinacol ester, 3-2-methoxyethylamine-1-carbonyl phenyl boronic acid pinacol ester, amtb047, 3-2-methoxyethylcarbamoyl phenylboronic acid, pinacol ester, n-2-methoxyethyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide
PubChem CID 46739043
IUPAC Name N-(2-methoxyethyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
SMILES COCCNC(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 93°C to 98°C
Quantity 1 g
Formula Weight 305.18
Percent Purity 97%
Chemical Name or Material 3-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.