accessibility menu, dialog, popup

UserName

BOOCH3CH3H3CCH3OHNOCH3

CAS RN 1073353-64-6

IUPAC Name: N-(2-methoxyethyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Synonyms: N-(2-methoxyethyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Molecular Weight (g/mol): 305.18
Molecular Formula: C16H24BNO4
InChi Key: GNJHHGGRMLSKNG-UHFFFAOYSA-N
SMILES: COCCNC(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results