missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Naphthalenyloxy)-1-phenyl-1-propanol, TRC

Product Code. 30480382
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30480382 10 mg
30472469 100 mg
30473577 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30480382 Supplier Toronto Research Chemicals Supplier No. TRCN35929010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 908291-72-5
Molecular Formula C19 H18 O2
Molecular Weight (g/mol) 278.34
Synonym α-[2-(1-Naphthalenyloxy)ethyl]benzenemethanol, 3-(1-Naphthalenyloxy)-1-phenylpropan-1-ol
IUPAC Name 3-naphthalen-1-yloxy-1-phenylpropan-1-ol
SMILES OC(CCOc1cccc2ccccc12)c3ccccc3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H18O2/c20-18(16-8-2-1-3-9-16)13-14-21-19-12-6-10-15-7-4-5-11-17(15)19/h1-12,18,20H,13-14H2
Formula Weight 278.1307
Chemical Name or Material 3-(1-Naphthalenyloxy)-1-phenyl-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.