CAS RN 908291-72-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(1-Naphthalenyloxy)-1-phenyl-1-propanol, TRC
CAS: 908291-72-5 Molecular Formula: C19 H18 O2 Molecular Weight (g/mol): 278.34 Synonym: α-[2-(1-Naphthalenyloxy)ethyl]benzenemethanol,3-(1-Naphthalenyloxy)-1-phenylpropan-1-ol IUPAC Name: 3-naphthalen-1-yloxy-1-phenylpropan-1-ol SMILES: OC(CCOc1cccc2ccccc12)c3ccccc3