missing translation for 'onlineSavingsMsg'
Learn More

(2R,5R)-1,6-Diphenylhexane-2,5-diamine Dihydrochloride, TRC

Product Code. 30382324
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382324 50 mg
30375435 500 mg
30383567 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382324 Supplier Toronto Research Chemicals Supplier No. TRCD49549850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1247119-31-8
Molecular Formula C18 H24 N2 . 2 Cl H
Molecular Weight (g/mol) 341.3184
Synonym 2,5-Hexanediamine, 1,6-diphenyl-, hydrochloride (1:2), (2R,5R)-, (R,R)-1,6-Diphenylhexane-2,5-diamine Dihydrochloride, (2R,5R)-1,6-Diphenylhexane-2,5-diamine Dihydrochloride
IUPAC Name (2R,5R)-1,6-diphenylhexane-2,5-diamine;dihydrochloride
SMILES Cl.Cl.N[C@H](CC[C@@H](N)Cc1ccccc1)Cc2ccccc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H24N2.2ClH/c19-17(13-15-7-3-1-4-8-15)11-12-18(20)14-16-9-5-2-6-10-16;;/h1-10,17-18H,11-14,19-20H2;2*1H/t17-,18-;;/m1../s1
Formula Weight 340.1473
Chemical Name or Material (2R,5R)-1,6-Diphenylhexane-2,5-diamine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.