CAS RN 1247119-31-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(2R,5R)-1,6-Diphenylhexane-2,5-diamine Dihydrochloride, TRC
CAS: 1247119-31-8 Molecular Formula: C18 H24 N2 . 2 Cl H Molecular Weight (g/mol): 341.3184 Synonym: 2,5-Hexanediamine, 1,6-diphenyl-, hydrochloride (1:2), (2R,5R)-,(R,R)-1,6-Diphenylhexane-2,5-diamine Dihydrochloride,(2R,5R)-1,6-Diphenylhexane-2,5-diamine Dihydrochloride IUPAC Name: (2R,5R)-1,6-diphenylhexane-2,5-diamine;dihydrochloride SMILES: Cl.Cl.N[C@H](CC[C@@H](N)Cc1ccccc1)Cc2ccccc2
2
(2R,5R)-1,6-Diphenylhexane-2,5-diamine Dihydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.