missing translation for 'onlineSavingsMsg'
Learn More

2H-Chromene-3-carboxylic acid, 90+%, Thermo Scientific™

Product Code. 10242782
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10242782 1 g 1g
10486462 250 mg 250mg
10460934 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10242782 Supplier Thermo Scientific Maybridge Supplier No. CC24101DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 22649-28-1
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
InChI Key DEBZQUFVQZPPLC-UHFFFAOYSA-N
Synonym 2h-1-benzopyran-3-carboxylic acid, 3-carboxy-2h-chromene, 2h-chromene-3-carboxylic, 2h-1-benzopyran-3-carboxylicacid
PubChem CID 322304
IUPAC Name 2H-chromene-3-carboxylic acid
SMILES C1C(=CC2=CC=CC=C2O1)C(=O)O

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 176.17
Percent Purity 90%
Chemical Name or Material 2H-Chromene-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.