accessibility menu, dialog, popup

UserName

OOO

CAS RN 22649-28-1

IUPAC Name: 2H-chromene-3-carboxylate
Synonyms: 2H-chromene-3-carboxylate
Molecular Weight (g/mol): 175.16
Molecular Formula: C10H7O3
InChi Key: DEBZQUFVQZPPLC-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC2=CC=CC=C2OC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results