missing translation for 'onlineSavingsMsg'
Learn More

(2E)-N-[(1S,2S)-2-Hydroxy-1-methyl-2-phenylethyl]-N-methyl-2-pentenamide, TRC

Product Code. 30395579
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395579 50 mg -
30391530 100 mg 100mg
30391128 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395579 Supplier Toronto Research Chemicals Supplier No. TRCH97283350MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 678991-33-8
Molecular Formula C15H21NO2
Molecular Weight (g/mol) 247.33
Synonym (E)-N-((1S,2S)-1-Hydroxy-1-phenylpropan-2-yl)-N-methylpent-2-enamide
IUPAC Name (E)-N-((1S,2S)-1-hydroxy-1-phenylpropan-2-yl)-N-methylpent-2-enamide
SMILES CC/C=C/C(N([C@@H](C)[C@@H](O)C1=CC=CC=C1)C)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H21NO2/c1-4-5-11-14(17)16(3)12(2)15(18)13-9-7-6-8-10-13/h5-12,15,18H,4H2,1-3H3/b11-5+/t12-,15+/m0/s1
Formula Weight 247.16
Chemical Name or Material (2E)-N-[(1S,2S)-2-Hydroxy-1-methyl-2-phenylethyl]-N-methyl-2-pentenamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.