CAS RN 678991-33-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2E)-N-[(1S,2S)-2-Hydroxy-1-methyl-2-phenylethyl]-N-methyl-2-pentenamide, TRC
CAS: 678991-33-8 Molecular Formula: C15H21NO2 Molecular Weight (g/mol): 247.33 Synonym: (E)-N-((1S,2S)-1-Hydroxy-1-phenylpropan-2-yl)-N-methylpent-2-enamide IUPAC Name: (E)-N-((1S,2S)-1-hydroxy-1-phenylpropan-2-yl)-N-methylpent-2-enamide SMILES: CC/C=C/C(N([C@@H](C)[C@@H](O)C1=CC=CC=C1)C)=O