missing translation for 'onlineSavingsMsg'
Learn More

(2E)-3-Fluoro-4-hydroxycinnamic Acid, TRC

Product Code. 30389039
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389039 50 mg
30401264 500 mg
30391356 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389039 Supplier Toronto Research Chemicals Supplier No. TRCF59922850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56926-74-0
Molecular Formula C9H7FO3
Molecular Weight (g/mol) 182.15
Synonym (2E)-3-(3-Fluoro-4-hydroxyphenyl)-2-propenoic Acid
SMILES OC(=O)\C=C\c1ccc(O)c(F)c1

Specifications

Quantity 50 mg
Formula Weight 182.0379
Chemical Name or Material (2E)-3-Fluoro-4-hydroxycinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.