Learn More
3M™ Polycarbonate Protective Safety Glasses, 2740 Series
Click to view available options
Lens Tint:
Amber
Clear
Gray
Conditionnement:
Each
Description
Additional features and style increase security, comfort, and wearer acceptance.
- Adjustable temple length to suit the individual
- Excellent impact resistance
- Polycarbonate lens provides reliable protection against specified levels of hazardous UV radiation
- Anti-scratch and anti-fog coating for increased durability and improved vision
- Optical Class 1 for prolonged use
- Ideal for use in many environments including agriculture, assembly work, automotive and bodyshops, construction, DIY, grinding, laboratories, light engineering, and woodworking
Spécification
Spécification
| Certifications/Compliance | EN 166:2001 |
| Lens Coating | Anti-fog, Scratch-resistant |
| Lens Tint | Gray |
| Product Type | Safety glasses; Safety Spectacles |
| For Use With (Application) | Construction, Metal fabrication, Forestry |
| Frame Color | Blue |
| No. per Pack | 1 Ea. |
| Temple Style | Adjustable length, comfortable fit |
| Lens Material | Polycarbonate |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu