missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SUPT16H Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals™ NBP3-30845-25ug

 View more versions of this product

Product Code. 30066781

  • £162.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SUPT16H Polyclonal antibody specifically detects SUPT16H in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SUPT16H
Polyclonal
Western Blot : 5-10 μg/mL, Immunoprecipitation
CDC68, Chromatin-specific transcription elongation factor 140 kDa subunit, facilitates chromatin remodeling 140 kDa subunit, Facilitates chromatin transcription complex subunit SPT16, FACT 140 kDa subunit, FACT complex subunit SPT16, FACT140, FACTp140, FACTP140FACT, FLJ10857, FLJ14010, FLJ34357, hSPT16, SPT16/CDC68, suppressor of Ty (S.cerevisiae) 16 homolog, suppressor of Ty 16 homolog (S. cerevisiae)
Recombinant protein consisting of amino acids 321-640 of the human SUPT16H protein conjugated to KLH.
25 μg
Primary
Human, Mouse
Purified
Immunoprecipitation, Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Ammonium sulfate precipitation
RUO
11198
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.