missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SUPT16H Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | SUPT16H |
|---|---|
| Dilution | Western Blot : 5-10 μg/mL, Immunoprecipitation |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066781
|
Novus Biologicals™
NBP3-30845-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067071
|
Novus Biologicals™
NBP3-30845-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SUPT16H Polyclonal antibody specifically detects SUPT16H in Human,Mouse samples. It is validated for Immunoprecipitation,Western BlotSpecifications
| SUPT16H | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| CDC68, Chromatin-specific transcription elongation factor 140 kDa subunit, facilitates chromatin remodeling 140 kDa subunit, Facilitates chromatin transcription complex subunit SPT16, FACT 140 kDa subunit, FACT complex subunit SPT16, FACT140, FACTp140, FACTP140FACT, FLJ10857, FLJ14010, FLJ34357, hSPT16, SPT16/CDC68, suppressor of Ty (S.cerevisiae) 16 homolog, suppressor of Ty 16 homolog (S. cerevisiae) | |
| Recombinant protein consisting of amino acids 321-640 of the human SUPT16H protein conjugated to KLH. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 5-10 μg/mL, Immunoprecipitation | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline | |
| 11198 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title