missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Marapsin/Pancreasin Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB19891-SP
This item is not returnable.
View return policy
Description
Marapsin/Pancreasin Monoclonal specifically detects Marapsin/Pancreasin in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| Marapsin/Pancreasin | |
| Monoclonal | |
| LYOPH | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q8BJR6 | |
| PRSS27 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Marapsin/Pancreasin Ala23-Thr290 Accession # Q8BJR6 | |
| 25 μg | |
| Primary | |
| Detects mouse Marapsin in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human Marapsin or recombinant mouse Prostasin was observed. | |
| Mouse | |
| Purified |
| ELISA | |
| 246406 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CAP-2, CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPN, MPNMarapsin, Pancreasin, protease, serine 27, PRSS27, serine protease 27 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 83886 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction