missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Marapsin/Pancreasin Antibody, R&D Systems™

Rat Monoclonal Antibody
£146.00 - £452.00
Specifications
| Antigen | Marapsin/Pancreasin |
|---|---|
| Clone | 246406 |
| Concentration | LYOPH |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18709628
![]() |
R&D Systems
MAB19891-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18190061
|
R&D Systems
MAB19891 |
500 μg |
£452.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Marapsin/Pancreasin Monoclonal specifically detects Marapsin/Pancreasin in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| Marapsin/Pancreasin | |
| LYOPH | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q8BJR6 | |
| 83886 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Marapsin/Pancreasin Ala23-Thr290 Accession # Q8BJR6 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects mouse Marapsin in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human Marapsin or recombinant mouse Prostasin was observed. |
| 246406 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CAP-2, CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPN, MPNMarapsin, Pancreasin, protease, serine 27, PRSS27, serine protease 27 | |
| PRSS27 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title