missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse FCRLB/FCRY Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB4868-SP
This item is not returnable.
View return policy
Description
FCRLB/FCRY Monoclonal specifically detects FCRLB/FCRY in Mouse samples. It is validated for Western Blot.
Specifications
| FCRLB/FCRY | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q5DRQ8 | |
| FCRLB | |
| Mouse myeloma cell line NS0-derived recombinant mouse FCRLB/FCRY Ala18-Ser427 Accession # Q5DRQ8 | |
| 25 μg | |
| Primary | |
| Detects mouse FCRLB/FCRY in direct ELISAs and Western blots. In Western blots, 75% cross-reactivity with recombinant human FCRLB is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 465905 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Fc receptor homolog expressed in B cells protein 2, Fc receptor like 2, Fc receptor-like and mucin-like 2, Fc receptor-like and mucin-like protein 2, Fc receptor-like B, Fc receptor-like protein 2, Fc receptor-related protein Y, FcRL2, FCRL2FCRLM2, FCRLM2, FCRLY, FCRY, FLJ31052, FREB2, FREB-2, FREB-2FcRY, RP11-474I16.6 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 127943 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction