missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse FCRLB/FCRY Antibody, R&D Systems™

Rat Monoclonal Antibody
£156.00 - £353.00
Specifications
| Antigen | FCRLB/FCRY |
|---|---|
| Clone | 465905 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18790299
![]() |
R&D Systems
MAB4868-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18112651
![]() |
R&D Systems
MAB4868 |
100 μg |
£353.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FCRLB/FCRY Monoclonal specifically detects FCRLB/FCRY in Mouse samples. It is validated for Western Blot.Specifications
| FCRLB/FCRY | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q5DRQ8 | |
| 127943 | |
| Mouse myeloma cell line NS0-derived recombinant mouse FCRLB/FCRY Ala18-Ser427 Accession # Q5DRQ8 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse FCRLB/FCRY in direct ELISAs and Western blots. In Western blots, 75% cross-reactivity with recombinant human FCRLB is observed. |
| 465905 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Fc receptor homolog expressed in B cells protein 2, Fc receptor like 2, Fc receptor-like and mucin-like 2, Fc receptor-like and mucin-like protein 2, Fc receptor-like B, Fc receptor-like protein 2, Fc receptor-related protein Y, FcRL2, FCRL2FCRLM2, FCRLM2, FCRLY, FCRY, FLJ31052, FREB2, FREB-2, FREB-2FcRY, RP11-474I16.6 | |
| FCRLB | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title