missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL2/JE/MCP-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB479R-SP
This item is not returnable.
View return policy
Description
CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CCL2/JE/MCP-1 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6347 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 123616R | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| P10148 | |
| Ccl2 | |
| E. coli-derived recombinant mouse CCL2 Gln24-Arg96 Accession # P10148 | |
| 25 μg | |
| Primary | |
| Detects mouse CCL2 in direct ELISAs. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction