missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL2/JE/MCP-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
£205.00 - £1740.00
Specifications
| Antigen | CCL2/JE/MCP-1 |
|---|---|
| Clone | 123616R |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18751199
![]() |
R&D Systems
MAB479R-SP |
25 μg |
£205.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18757188
![]() |
R&D Systems
MAB479R-100 |
100 μg |
£438.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18142641
|
R&D Systems
MAB479R-500 |
500 ug |
£1740.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18148920
|
R&D Systems
MAB479R-01M |
1 mg |
£1460.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Mouse samples. It is validated for Flow Cytometry.Specifications
| CCL2/JE/MCP-1 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| P10148 | |
| 6347 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Detects mouse CCL2 in direct ELISAs. |
| 123616R | |
| Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF | |
| Ccl2 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title