missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human POU2F3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11625-SP
This item is not returnable.
View return policy
Description
POU2F3 Monoclonal antibody specifically detects POU2F3 in Human samples. It is validated for Immunohistochemistry
Specifications
| POU2F3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| Epoc-1, MGC126698, oct-11, OCT11, OCT11FLJ40063, Octamer-binding protein 11, Octamer-binding transcription factor 11, OTF11, OTF-11, PLA1, PLA-1, POU class 2 homeobox 3, POU domain class 2, transcription factor 3, POU domain, class 2, transcription factor 3, POU transcription factor, Skn-1a, Transcription factor PLA-1, Transcription factor Skn-1 | |
| Synthetic Peptide, Accession # Q9UKI9 | |
| 25 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry | |
| 1091214 | |
| Immunohistochemistry 3-25 μg/mL | |
| Q9UKI9 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 25833 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction