missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human POU2F3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
£218.00 - £481.00
Specifications
| Antigen | POU2F3 |
|---|---|
| Clone | 1091214 |
| Dilution | Immunohistochemistry 3-25 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279912
|
R&D Systems
MAB11625-SP |
25 μg |
£218.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279890
|
R&D Systems
MAB11625-100 |
100 μg |
£481.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
POU2F3 Monoclonal antibody specifically detects POU2F3 in Human samples. It is validated for ImmunohistochemistrySpecifications
| POU2F3 | |
| Immunohistochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| Epoc-1, MGC126698, oct-11, OCT11, OCT11FLJ40063, Octamer-binding protein 11, Octamer-binding transcription factor 11, OTF11, OTF-11, PLA1, PLA-1, POU class 2 homeobox 3, POU domain class 2, transcription factor 3, POU domain, class 2, transcription factor 3, POU transcription factor, Skn-1a, Transcription factor PLA-1, Transcription factor Skn-1 | |
| Synthetic Peptide, Accession # Q9UKI9 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1091214 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UKI9 | |
| 25833 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title