Learn More
Human Desmin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB11607-SP
 View more versions of this product
Product Code. 30279906
  • £218.00 / 25µg
Description
Desmin Monoclonal antibody specifically detects Desmin in Human samples. It is validated for Immunocytochemistry,Western Blot,Simple Western,Immunohistochemistry,Multiplex Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Desmin
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
CMD1I, CMD1IFLJ41013, CSM1, CSM2, DES, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein
E. coli derived recombinant human Desmin, Accession # P17661
25 μg
Primary
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution.
IgG2a
Immunocytochemistry, Western Blot, Simple Western, Immunohistochemistry, Multiplex
1089115
Western Blot 1 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL, Multiplex Immunofluorescence 1 μg/mL
P17661
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
1674
Human
Purified