missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Desmin Antibody, R&D Systems™
Mouse Monoclonal Antibody
£218.00 - £490.00
Specifications
| Antigen | Desmin |
|---|---|
| Clone | 1089115 |
| Dilution | Western Blot 1 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL, Multiplex Immunofluorescence 1 μg/mL |
| Applications | Immunocytochemistry, Western Blot, Simple Western, Immunohistochemistry, Multiplex |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279906
|
R&D Systems
MAB11607-SP |
25 μg |
£218.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279885
|
R&D Systems
MAB11607-100 |
100 μg |
£490.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Desmin Monoclonal antibody specifically detects Desmin in Human samples. It is validated for Immunocytochemistry,Western Blot,Simple Western,Immunohistochemistry,Multiplex ImmunofluorescenceSpecifications
| Desmin | |
| Western Blot 1 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL, Multiplex Immunofluorescence 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| CMD1I, CMD1IFLJ41013, CSM1, CSM2, DES, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
| E. coli derived recombinant human Desmin, Accession # P17661 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1089115 | |
| Immunocytochemistry, Western Blot, Simple Western, Immunohistochemistry, Multiplex | |
| Unconjugated | |
| Mouse | |
| Human | |
| P17661 | |
| 1674 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title