missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Factor H-related 1/CFHR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB4247-SP
This item is not returnable.
View return policy
Description
Complement Factor H-related 1/CFHR1/CFHL1 Monoclonal specifically detects Complement Factor H-related 1/CFHR1/CFHL1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Complement Factor H-related 1/CFHR1/CFHL1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q03591 | |
| CFHR1 | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Factor H-related 1/CFHR1 Glu19-Ala328 Accession # Q03591 | |
| 25 μg | |
| Primary | |
| Detects human Complement Factor H-related 1/CFHR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CFHR2 or rhCFHR5 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 442127 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BTA, CFHL1FHR-1, CFHLCFHR1P, CFHR1, complement factor H-related 1, Complement Factor Hrelated 1, complement factor H-related 1 pseudogene, complement factor H-related protein 1, FHR1, FHR1CFHL1P, H factor (complement)-like 1, H factor (complement)-like 2, H factor-like protein 1, H36-1, H36-2, H-factor-like 1, HFL1H36, HFL2, MGC104329 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3078 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction