missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Plastic Lock Bacti Caps

Product Code. 11532662 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11532662 Green Pack of 50
11522662 Blue Pack of 50
10428392 Blue -
11502662 Red Pack of 50
11592652 White Pack of 50
11512662 Yellow Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11532662
Only null left
Add to basket
Add to basket

Made from chemically resistant virgin plastic, Bacti-Caps™ provide a firm, positive culture tube closure allowing a controlled gas interchange. They are ideal for use in science, research, and other laboratories.

  • Bacti-Caps™  for test tubes without edge
  • Provide a firm, positive culture tube closure allowing a controlled gas interchange, thus proving a more practical alternative to cotton wool plugs
  • Suitable for all standard test tube sizes, they fit rimless and plastic tubes with an outside diameter of 13mm, 16mm, 19mm, 25mm and 38mm 
  • The three flexible fins molded on the inner wall of each cap ensure a positive fit even when minor variations in tube size occur
  • Bacti-Caps™ are made from chemically resistant virgin plastic so they are unaffected by culture media, most acids, alkalis, alcohols and esters
  • Improved culture growth
  • Color coded
  • Controlled gas interchange

Specifications

Product Type Lock Bacti Cap
Autoclavable Autoclavable
Material Plastic
Color Green
Quantity 100 Pack
Size (Metric) 25 mm
For Use With (Equipment) Test Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.