missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NPRC/NPR3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10233-SP
This item is not returnable.
View return policy
Description
Natriuretic Peptide Receptor C Monoclonal antibody specifically detects Natriuretic Peptide Receptor C in Human samples. It is validated for Immunohistochemistry
Specifications
| Natriuretic Peptide Receptor C | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| ANP-C, ANPR-C, ANPRCFLJ14054, Atrial natriuretic peptide clearance receptor, atrial natriuretic peptide receptor 3, Atrial natriuretic peptide receptor type C, C5orf23, chromosome 5 open reading frame 23, GUCY2B, lgj, natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptidereceptor C), NPR3, NPRC, NPR-C, stri | |
| Human embryonic kidney cell, HEK293-derived human NPRC/NPR3, Met1-Glu481, Accession # P17342 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
| Immunohistochemistry | |
| 1060839 | |
| Immunohistochemistry 5-15 ug/mL | |
| P17342 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4883 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction