missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NPRC/NPR3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£174.00 - £520.00
Specifications
| Antigen | Natriuretic Peptide Receptor C |
|---|---|
| Clone | 1060839 |
| Dilution | Immunohistochemistry 5-15 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18716115
![]() |
R&D Systems
MAB10233-100 |
100 μg |
£520.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18705895
![]() |
R&D Systems
MAB10233-SP |
25 μg |
£174.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Natriuretic Peptide Receptor C Monoclonal antibody specifically detects Natriuretic Peptide Receptor C in Human samples. It is validated for ImmunohistochemistrySpecifications
| Natriuretic Peptide Receptor C | |
| Immunohistochemistry 5-15 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| ANP-C, ANPR-C, ANPRCFLJ14054, Atrial natriuretic peptide clearance receptor, atrial natriuretic peptide receptor 3, Atrial natriuretic peptide receptor type C, C5orf23, chromosome 5 open reading frame 23, GUCY2B, lgj, natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptidereceptor C), NPR3, NPRC, NPR-C, stri | |
| Human embryonic kidney cell, HEK293-derived human NPRC/NPR3, Met1-Glu481, Accession # P17342 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1060839 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P17342 | |
| 4883 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title