Learn More
Human/Mouse/Rat MIOS Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB9289-SP
 View more versions of this product
Product Code. 18786789
  • £160.00 / 25µg
Description
MIOS Monoclonal specifically detects MIOS in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MIOS
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
FLJ20323, MIO, missing oocyte, meiosis regulator, homolog (Drosophila), WD repeat-containing protein mio
Human MIOS synthetic peptide Accession # Q9NXC5
25 μg
Primary
Detects human MIOS in direct ELISAs and Western blots.
Human, Mouse, Rat
Purified
Western Blot
955413
Western Blot 2 ug/mL
Q9NXC5
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
54468
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1