missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat MIOS Antibody, R&D Systems™

Mouse Monoclonal Antibody
£160.00 - £409.00
Specifications
| Antigen | MIOS |
|---|---|
| Clone | 955413 |
| Dilution | Western Blot 2 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18786789
![]() |
R&D Systems
MAB9289-SP |
25 μg |
£160.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18735838
![]() |
R&D Systems
MAB9289-100 |
100 μg |
£409.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MIOS Monoclonal specifically detects MIOS in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| MIOS | |
| Western Blot 2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| FLJ20323, MIO, missing oocyte, meiosis regulator, homolog (Drosophila), WD repeat-containing protein mio | |
| Human MIOS synthetic peptide Accession # Q9NXC5 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human MIOS in direct ELISAs and Western blots. |
| 955413 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q9NXC5 | |
| 54468 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title