missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Galectin-9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB20451-SP
This item is not returnable.
View return policy
Description
Galectin-9 Monoclonal specifically detects Galectin-9 in Human samples. It is validated for ELISA.
Specifications
| Galectin-9 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| ecalectin, gal-9, GAL9, galectin 9, galectin-9, Galectin9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein | |
| Human embryonic kidney cell, HEK293derived human Galectin-9, Met1-Thr323, Accession # NP_002299 | |
| 25 μg | |
| Primary | |
| Detects human Galectin-9 in direct ELISAs. | |
| Human | |
| Purified |
| ELISA | |
| 1015223 | |
| ELISA | |
| NP_002299 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3965 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction