missing translation for 'onlineSavingsMsg'
Learn More

Human Galectin-9 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£164.00 - £408.00

Specifications

Antigen Galectin-9
Clone 1015223
Dilution ELISA
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18700287
GREENER_CHOICE
View Documents
R&D Systems
MAB20451-SP
25 μg
£164.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18733477
GREENER_CHOICE
View Documents
R&D Systems
MAB20451-100
100 μg
£408.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Galectin-9 Monoclonal specifically detects Galectin-9 in Human samples. It is validated for ELISA.
Specifications

Specifications

Galectin-9
ELISA
Monoclonal
Purified
RUO
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
ecalectin, gal-9, GAL9, galectin 9, galectin-9, Galectin9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein
Human embryonic kidney cell, HEK293derived human Galectin-9, Met1-Thr323, Accession # NP_002299
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Galectin-9 in direct ELISAs.
1015223
ELISA
Unconjugated
Mouse
Human
NP_002299
3965
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.