missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fibronectin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB19182-SP
This item is not returnable.
View return policy
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| Fibronectin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
| E.coli-derived recombinant human Fibronectin Asn631-Pro705 Accession # P02751 | |
| 25 μg | |
| Primary | |
| Detects human Fibronectin in direct ELISAs and Western blots. | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 960642 | |
| Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| P02751 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 2335 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction