missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fibronectin Antibody, R&D Systems™

Mouse Monoclonal Antibody
£177.00 - £457.00
Specifications
| Antigen | Fibronectin |
|---|---|
| Clone | 960642 |
| Dilution | Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 3-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18714899
![]() |
R&D Systems
MAB19182-SP |
25 μg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18779777
![]() |
R&D Systems
MAB19182-100 |
100 μg |
£457.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
| Fibronectin | |
| Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P02751 | |
| 2335 | |
| IgG1 | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 960642 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
| E.coli-derived recombinant human Fibronectin Asn631-Pro705 Accession # P02751 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Fibronectin in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title