missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 24 Listeria Enrichment Broth (LEB) Buffer Supplement

Product Code. 13245549
Change view
Click to view available options
Fill Weight:
113.0-114.7g
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight unitSize
13245549 113.0-114.7g Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13245549 Supplier Thermo Scientific™ Supplier No. BO1204M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optimize growing conditions for all Listeria species prior to molecular assay with Thermo Scientific™ 24 LEB Buffer Supplement, added to our range of complementary media.

Enrich Listeria monocytogenes and Listeria spp. from food and environmental samples prior to molecular assay with the Thermo Scientific™ 24 Listeria Enrichment Broth (LEB) Buffer Supplement. The supplement is part of the 24 LEB range which includes complementary media and provides optimal growing conditions for all Listeria species for PCR applications. It is specifically designed for use with PCR Systems, such as Thermo Scientific™ SureTect™ molecular assays for Listeria.

The 24 LEB product range provides rapid growth of all Listeria species and has been validated with Thermo Scientific™ SureTect™ molecular assays for Listeria to provide results in 24 hours from food and environmental samples.

Achieve optimal growing conditions for all Listeria species with 24 LEB Buffer Supplement added to 24 LEB Broth (BO1205S) and 24 LEB Dehydrated Media Complete Base (CM1154).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless, slight haze
Content And Storage 2 to 25°C
Format 100mL in 125mL PET Sirop
Manufacturing Origin United Kingdom
Product Type Microbiology Media
Fill Weight 113.0-114.7g
Quantity 10/Pk.

Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.