missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 24 LEB Buffer Supplement
Description
The 24 LEB product range provides rapid growth of all Listeria species and has been validated with Thermo Scientific™ SureTectTM molecular assays for Listeria to provide results in 24 hours from food and environmental samples.
- The 24 LEB product range provides rapid growth of all Listeria species and has been validated with Thermo Scientific™ SureTectTM molecular assays for Listeria to provide results in 24 hours from food and environmental samples
Specifications
Specifications
| Color | Colorless, slight haze |
| Content And Storage | 2 to 25°C |
| Format | Universal bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Solution |
| Fill Weight | 11.3-11.6g |
| Packaging Type | Twenty-four universal bottles with screw cap closures in a box. |
| Quantity | 24/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?