Learn More
Invitrogen™ MLKL Recombinant Rabbit Monoclonal Antibody (23GB2935), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553519
 View more versions of this product
Product Code. 30088087
  • £179.00 / 20µL
Description
The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MLKL
Recombinant Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8NB16
Mlkl
A synthesized peptide derived from MLKL (1-200AA).
20 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB2935
Unconjugated
Mlkl
9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like
Rabbit
Affinity chromatography
RUO
197259
-20°C