Learn More
Invitrogen™ GNE Recombinant Rabbit Monoclonal Antibody (23GB5560), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552881
 View more versions of this product
Product Code. 30089198
  • £162.00 / 20µL
Description
GNE is a bifunctional enzyme that initiates and regulates the biosynthesis of N-acetylneuraminic acid (NeuAc), a precursor of sialic acids. It is a rate-limiting enzyme in the sialic acid biosynthetic pathway. Sialic acid modification of cell surface molecules is crucial for their function in many biologic processes, including cell adhesion and signal transduction. Differential sialylation of cell surface molecules is also implicated in the tumorigenicity and metastatic behavior of malignant cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GNE
Recombinant Monoclonal
Unconjugated
GNE
2310066H07Rik; bifunctional UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase; DMRV; GLCNE; glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase; GNE; IBM2; ManAc kinase; N-acetylmannosamine kinase; N-acylmannosamine kinase; NM; Uae1; UDP-GlcNAc-2-epimerase; UDP-GlcNAc-2-epimerase/ManAc kinase; UDP-N-acetylglucosamine 2-epimerase (hydrolyzing); UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase; UDP-N-acetylglucosamine-2-epimerase/N-acetylmannosamine kinase; Uridine diphosphate-N-acetylglucosamine-2-epimerase
Rabbit
Affinity chromatography
RUO
10020, 50798
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB5560
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q91WG8, Q9Y223
GNE
A synthesized peptide derived from human GNE (701-722AA).
20 μL
Primary
Human, Mouse