missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNE Recombinant Rabbit Monoclonal Antibody (23GB5560), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | GNE |
|---|---|
| Clone | 23GB5560 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088864
![]() |
Invitrogen™
MA552880 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089198
|
Invitrogen™
MA552881 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GNE Recombinant Monoclonal antibody specifically detects GNE in Human, Mouse samples. It is validated for Western BlotSpecifications
| GNE | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q91WG8, Q9Y223 | |
| 10020, 50798 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5560 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2310066H07Rik; bifunctional UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase; DMRV; GLCNE; glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase; GNE; IBM2; ManAc kinase; N-acetylmannosamine kinase; N-acylmannosamine kinase; NM; Uae1; UDP-GlcNAc-2-epimerase; UDP-GlcNAc-2-epimerase/ManAc kinase; UDP-N-acetylglucosamine 2-epimerase (hydrolyzing); UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase; UDP-N-acetylglucosamine-2-epimerase/N-acetylmannosamine kinase; Uridine diphosphate-N-acetylglucosamine-2-epimerase | |
| A synthesized peptide derived from human GNE (701-722AA). | |
| Primary | |
| -20°C | |
| GNE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title