missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHD1L Recombinant Rabbit Monoclonal Antibody (23GB3770), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552571
This item is not returnable.
View return policy
Description
In response to DNA strand breaks, chromatin adopts a relaxed structure due to the addition of poly(ADP-ribose) (PAR) to chromatin proteins by PARP enzymes (see PARP1; MIM 173870), and this relaxation facilitates the repair of DNA damage. CHD1L interacts with PAR and has a role in chromatin relaxation following DNA damage (Ahel et al., 2009 [PubMed 19661379]).
Specifications
| CHD1L | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CHD1L | |
| 4432404A22Rik; ALC1; amplified in liver cancer 1; amplified in liver cancer protein 1; CHD1L; CHDL; chromodomain helicase DNA binding protein 1 like; chromodomain helicase DNA binding protein 1-like; chromodomain-helicase-DNA-binding protein 1-like; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 1-like; Snf2p; Unknown (protein for MGC:128792) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9557 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3770 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86WJ1 | |
| CHD1L | |
| A synthetic peptide of human CHD1L (700-800AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction