missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHD1L Recombinant Rabbit Monoclonal Antibody (23GB3770), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | CHD1L |
|---|---|
| Clone | 23GB3770 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089851
![]() |
Invitrogen™
MA552570 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087528
|
Invitrogen™
MA552571 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHD1L Recombinant Monoclonal antibody specifically detects CHD1L in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| CHD1L | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q86WJ1 | |
| 9557 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3770 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 4432404A22Rik; ALC1; amplified in liver cancer 1; amplified in liver cancer protein 1; CHD1L; CHDL; chromodomain helicase DNA binding protein 1 like; chromodomain helicase DNA binding protein 1-like; chromodomain-helicase-DNA-binding protein 1-like; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 1-like; Snf2p; Unknown (protein for MGC:128792) | |
| A synthetic peptide of human CHD1L (700-800AA). | |
| Primary | |
| -20°C | |
| CHD1L |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title