missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IMP2/IGF2BP2 Antibody (PCRP-IGF2BP2-1F9), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-26764-20ug

 View more versions of this product

Product Code. 30043294

  • £178.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IMP2/IGF2BP2 Monoclonal antibody specifically detects IMP2/IGF2BP2 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IMP2/IGF2BP2
Monoclonal
0.2 mg/mL
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Flow Cytometry
PCRP-IGF2BP2-1F9
Unconjugated
Hepatocellular carcinoma autoantigen p62, IGF-II mRNA-binding protein 2, IMP-2IGF2 mRNA-binding protein 2, IMP2VICKZ family member 2, insulin-like growth factor 2 mRNA binding protein 2, insulin-like growth factor 2 mRNA-binding protein 2, p62, VICKZ2
Recombinant full-length human IMP2/IGF2BP2 protein
20 μg
Cancer, Chromatin Research, Immune System Diseases, Immunology, Transcription Factors and Regulators
10644
Store at 4°C.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.