missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IMP2/IGF2BP2 Antibody (PCRP-IGF2BP2-1F9), Novus Biologicals™
Mouse Monoclonal Antibody
£178.00 - £351.00
Specifications
| Antigen | IMP2/IGF2BP2 |
|---|---|
| Clone | PCRP-IGF2BP2-1F9 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043294
|
Novus Biologicals™
NBP3-26764-20ug |
20 μg |
£178.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043070
|
Novus Biologicals™
NBP3-26764-100ug |
100 μg |
£351.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IMP2/IGF2BP2 Monoclonal antibody specifically detects IMP2/IGF2BP2 in Human samples. It is validated for Flow CytometrySpecifications
| IMP2/IGF2BP2 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Hepatocellular carcinoma autoantigen p62, IGF-II mRNA-binding protein 2, IMP-2IGF2 mRNA-binding protein 2, IMP2VICKZ family member 2, insulin-like growth factor 2 mRNA binding protein 2, insulin-like growth factor 2 mRNA-binding protein 2, p62, VICKZ2 | |
| Recombinant full-length human IMP2/IGF2BP2 protein | |
| Primary | |
| Store at 4°C. |
| PCRP-IGF2BP2-1F9 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Cancer, Chromatin Research, Immune System Diseases, Immunology, Transcription Factors and Regulators | |
| 10 mM PBS with 0.05% BSA | |
| 10644 | |
| IgG | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title