missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 2000μL Polypropylene 96-Well Deep Well PCR Plates

Combine a new well design with a sophisticated color coding system for ease of reading and orientation.The well geometry of Eppendorf™ 2000μL Polypropylene 96-Well Deep Well PCR Plates allows high speed pipetting without splashing, excellent mixing and high speed centrifugation, which accelerates your manual and automated work processes.

Brand:  Eppendorf™ 0030505301

Code : 96

Additional Details : Weight : 8.55000kg

 View more versions of this product

Product Code. 10245983

  • £558.20 / Pack of 80


Estimated Shipment
20-02-2018
Add to basket

Description

Description

  • Fulfill all current SBS, IVD, EN ISO 9001/2/3 and EN 13485 guidelines.
  • Recovermax - concaved round bottom well shape with smoothed edges
  • Optitrack - color framed plates with alphanumeric labelling
  • High chemical resistance
  • G-safe - highest centrifugation stability
  • LoBind - free of surface coating e.g., silicone
  • Free from DNA, DNase, RNase and PCR* inhibitor (PCR* clean)
  • LoBind Protein - minimum protein loss (less than 3% BSA 1μg/mL)
  • LoBind DNA - less than 1% loss of DNA/RNA at low concentrations and critical conditions (high salt, short DNA fragments)
  • Autoclavable
  • Optionally available with barcode
Specifications

Specifications

96
2000μL
Videos
Documentation

Technical Documents